Skip to Content
Merck
All Photos(1)

Key Documents

566327

Sigma-Aldrich

SIRT1/2 Inhibitor VII

The SIRT1/2 Inhibitor VII, also referenced under CAS 143034-06-4, controls the biological activity of SIRT1/2. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT1/2 Inhibitor VII, 5-(2-Naphthylidine)-2-thiobarbituric acid, SIRT1 Inhibitor V, SIRT2 Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O2S
CAS Number:
Molecular Weight:
282.32
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A thiobarbiturate compound that acts as an inhibitor against sirtuins (IC50 = 13.2 & 9.1 µM using human SIRT1 & SIRT2, respectively), the NAD+-dependent class III histone deacetylases (HDACs). Computer-aided molecular structural anaylsis reveals that both hydrogen bonds and van der Waals type of interactions contribute to the successful docking of the inhibitor into the nicotinamde subpocket of SIRT2 with the bulky naphthyl group being placed directly into the acetyllysine substrate channel.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Uciechowska, U., et al. 2008. ChemMedChem3, 1965.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service