444224
MMP-2/MMP-9 Substrate II
An exceptional thiopeptide that is used as a substrate for the rapid, sensitive, and continuous spectrophotometric assay of Type IV collagenases, MMP-2 (gelatinase A), and MMP-9 (gelatinase B).
Synonym(s):
MMP-2/MMP-9 Substrate II, Ac-Pro-Leu-Gly-(2-mercapto-4-methylpentanoyl)-Leu-Gly-OEt, Gelatinase A/Gelatinase B Substrate II
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
water: 1 mg/mL
50% acetic acid: 10 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
Biochem/physiol Actions
Substrate for the rapid, sensitive, and continuous spectrophotometric assay of Type IV collagenases, MMP-2 (gelatinase A), and MMP-9 (gelatinase B).
Warning
Sequence
Reconstitution
Other Notes
Weingarten, H., et al. 1985. Biochemistry 24, 6730.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service