Skip to Content
Merck
All Photos(3)

Key Documents

773840

Sigma-Aldrich

Carbon nanotube, multi-walled

≥98% carbon basis, O.D. × I.D. × L 10 nm ± 1 nm × 4.5 nm ± 0.5 nm × 3-~6 μm, TEM, avg. no. of layers, 6 ‑ 8

Synonym(s):

SMW200, MWCNT, MWNT, Multiwall carbon nanotube

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103

Assay

≥98% carbon basis

form

powder

feature

avg. no. of layers 6 ‑ 8

O.D. × I.D. × L

10 nm ± 1 nm × 4.5 nm ± 0.5 nm × 3-~6 μm, TEM

surface area

280-350 m2/g , BET

mp

3652-3697 °C (lit.)

density

~2.1 g/mL at 25 °C (lit.)

bulk density

0.068 g/cm3

Looking for similar products? Visit Product Comparison Guide

General description

Number of Walls: 6-8 tube walls (typically). Multi walled carbon nanotubes (MWNTs, CNTs) were prepared by chemical vapor deposition (CVD) using cobalt and molybdenum as catalysts (CoMoCAT).In chemical vapor deposition (CVD), a volatile precursor undergoes thermal decomposition at elevated temperatures to form a solid deposit on a substrate. Compared to other techniques CVD produces greater yield of carbon nanotubes. Morphology and structure of the nanotubes can be easily tuned using CVD technique. The CNTs formed can be formed in bundles over a catalyst support or in patterns over substrates.1

Application

Carbon nanotube, multi-walled (MWNT) belongs to the class of carbonaceous materials with excellent physiochemical, thermo-mechanical and electrochemical properties. This material can be used in a variety of sustainable energy applications such as solar cells, photocatalysis, biosensor, gas sensor, supercapacitor and as a filler that acts as a reinforcement to improve the mechanical property of composites.
New generation of specialty multi-walled carbon nanotubes enable polymer composites with higher conductivity;easier to disperse and mechanical properties for advanced applications like conductive polymers or Li-ion battery cathodes.

Physical properties

Aspect ratio: > 350-550 (determined by AFM)

Preparation Note

CoMoCAT Catalytic Chemical Vapor Deposition (CVD) Method

Legal Information

Product of SouthWest NanoTechnologies;Inc. CoMoCAT® is a Registered Trademark of SouthWest NanoTechnologies;Inc.
 
SWeNT is a registered trademark of Southwest NanoTechnologies;Inc.
CoMoCAT is a trademark of Chasm Advanced Materials

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Jin et al.
Electrochemical Communications, 10, 10537-11540 (2008)
K. Sheem et al.
Journal of Power Sources, 158, 1425-1430 (2006)
Olli Pitkänen et al.
Nanotechnology, 31(49), 495403-495403 (2020-09-26)
There is ever increasing demand for flexible energy storage devices due to the development of wearable electronics and other small electronic devices. The electrode flexibility is best provided by a special set of nanomaterials, but the required methodology typically consists
S.K. Martha et al.
Journal of Power Sources, 189, 288-296 (2009)
Claudia Angélica Ramírez-Herrera et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-02-13)
The demand for multifunctional requirements in aerospace, military, automobile, sports, and energy applications has encouraged the investigation of new composite materials. This study focuses on the development of multiwall carbon nanotube (MWCNT) filled polypropylene composites and carbon nanofiber composite mats.

Articles

Improved multi-walled carbon nanotubes (MWNTs) have been recently developed by CoMoCAT® technique and adopted in a significant number of applications, including batteries, water filter membranes, and electric or thermal conducting polymer composites.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service