Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200471

Sigma-Aldrich

Anti-SND1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-TDRD11, Anti-staphylococcal nuclease and tudor domain containing 1, p100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~102 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1-2 μg using lysates of rat NRK cells.
indirect immunofluorescence: 2.5-5.0 μg/mL using human HeLa cells
western blot: 0.25-0.5 μg/mL using whole extracts of mouse NIH-3T3 cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SND1(27044)
mouse ... Snd1(56463)
rat ... Snd1(64635)

General description

Staphylococcal nuclease and tudor domain containing 1 (SND1) protein contains five staphylococcal nuclease-like domains and a tudor-like domain. SND1 is a component of the RNA-induced silencing complex (RISC). This gene is highly conserved from yeast to human.

Immunogen

peptide corresponding to the C-terminal region of human SND1 (GeneID: 27044), conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Application

Anti-SND1 (C-terminal) antibody produced in rabbit has been used in immunoblotting, immunoprecipitation and immunofluorescence.

Biochem/physiol Actions

Staphylococcal nuclease and tudor domain containing 1 (SND1) has been shown to be involved in the regulation of transcription and RNA biogenesis. It is known to interact with nucleic acids and proteins. SND1 promotes the breakdown of hyper-edited inosine-containing miRNA precursors. It modulates miRNA transcription and translation through RNA editing by ADAR (adenosine deaminase acting on RNA). Increased SND1 expression is seen in human colon cancer tissues and cell lines.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and functional insights into human Tudor-SN, a key component linking RNA interference and editing
Li CL, et al.
Nucleic Acids Research, 36(11), 3579-3589 (2008)
Naoto Tsuchiya et al.
Cancer research, 67(19), 9568-9576 (2007-10-03)
Colon cancers have been shown to develop after accumulation of multiple genetic and epigenetic alterations with changes in global gene expression profiles, contributing to the establishment of widely diverse phenotypes. Transcriptional and posttranscriptional regulation of gene expression by small RNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service