39905
Lectin from Phytolacca americana-Atto 488 conjugate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
fluorescence
λex 501 nm; λem 523 nm in PBS
Analysis Note
free of unconjugated dye
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Breaking the Diffraction Barrier in Fluorescence Microscopy by Optical Shelving.
Physical Review Letters, 98(21) (2007)
Dual color localization microscopy of cellular nanostructures.
Biotechnology Journal, 4(6), 927-938 (2009)
Super-Resolution Imaging of Fluorophore-Labeled DNA Bound to Gold Nanoparticles. A Single-Molecule, Single-Particle Approach.
The Journal of Physical Chemistry C, 120(2), 803?815-803?815 (2016)
Effects of Tuning Fluorophore Density, Identity, and Spacing on Reconstructed Images in Super-Resolution Imaging of Fluorophore-Labeled Gold Nanorods.
The Journal of Physical Chemistry, 119(50), 28099?28110-28099?28110 (2015)
Dynamic Superresolution Imaging of Endogenous Proteins on Living Cells at Ultra-High Density.
Biophysical Journal, 99(4), 1303-1310 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service