Skip to Content
Merck
All Photos(1)

Key Documents

15-1060

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

SAJ first grade, ≥98.0%

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ first grade
form:
crystalline

grade

SAJ first grade

Assay

≥98.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

1 (25 °C, 482.2 g/L)

mp

39-41 °C (lit.)

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Puntarulo et al.
Archives of biochemistry and biophysics, 264(2), 482-491 (1988-08-01)
The interaction of microsomes with iron and NADPH to generate active oxygen radicals was determined by assaying for low level chemiluminescence. The ability of several ferric complexes to catalyze light emission was compared to their effect on microsomal lipid peroxidation
Luigi Brunetti et al.
Neurobiology of aging, 25(1), 125-129 (2003-12-17)
In order to investigate whether amyloid beta-peptide-induced oxidative damage in the brain could be related to aging, we studied the release of 8-iso-prostaglandin (PG)F2alpha, a stable marker of cellular oxidative stress, in brain synaptosomes from Wistar rats of different ages
R Săndulescu et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 951-958 (1996-06-01)
The method is based upon the reaction between fluoride ions and the coloured complex of Fe(III) with methyl salicylate to form the stable, colourless hexaflouride complex of iron. The conditions of the method (pH, time and combination ratio) were studied
Jun-Beum Lee et al.
Bioscience, biotechnology, and biochemistry, 68(3), 495-500 (2004-04-02)
Efficiency was examined of microencapsulating L-ascorbic acid by polyglycerol monostearate (PGMS), and changes in the chemical and sensorial aspects of L-ascorbic acid and/or iron-fortified milk during storage were evaluated. The selected core materials were ferric ammonium sulfate and L-ascorbic acid.
M X Yang et al.
Archives of biochemistry and biophysics, 324(2), 282-292 (1995-12-20)
The NADH-dependent microsomal electron transfer system consists of NADH-cytochrome b5 reductase and cytochrome b5, which donates reducing equivalents to fatty acyl desaturase, cytochrome P450, and other reactions. A study was carried out to investigate the interaction of NADH with several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service