Skip to Content
Merck
All Photos(3)

Key Documents

ABE353

Sigma-Aldrich

Anti-dimethyl Histone H2B Antibody (Pro1)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2B type 1-C/E/F/G/, H2Bme2P1, Histone H2B.1 A, Histone H2B.a, H2B/a, Histone H2B.g, H2B/g, Histone H2B.h, Histone H2B.k, H2B.l, H2B/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Pro1)

Gene Information

General description

Histone H2B is one of four histones that form a core octameric structure required for the packaging of DNA into nucleosomes, the most basic level of chromatin arrangement. Histones undergo a number of post-translational modifications (PTM) at their N- and C-terminal tails which can influence chromatin structure and stability; and consequently, transcription, and DNA replication and repair. Acetylation is a reversible PTM that involves the transfer of acetyl groups to N-terminal lysine residues by a family of histone acetyltransferase enzymes, including GNAT, Gcn5, and p300/CBP. Histone acetylation reduces the electrostatic attractions between DNA and histones, which destabilizes chromatin structure and provide transcriptional complexes, for example, with access to DNA, potentially resulting in increased gene transcription. Lysine 120 of H2B (H2BK120) undergoes acetylation (by p300/CBP) and ubiquitination. Acetylation occurs first and is required for subsequent mono-ubiquitination. H2BK120 may function as a marker of “pre-active” chromatin, and is part of a complex regulatory system involving multiple histone modifications.

Specificity

Recognizes Histone H2B dimethylated at Proline 1.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of Human dimethyl Histone H2B methylated at Pro1.

Application

Dot Blot Specificity Analysis:
Peptide containing various unmodified and modified Histone peptides were probed with Anti-dimethyl Histone H2B (Pro1) antibody at 0.1 ug/mL.

Immunocytochemistry Analysis:
A 10 µg/mL dilution from a representative lot detected Histone H2B dimethylated at proline 1 in A431, HeLa, and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology
This Anti-dimethyl Histone H2B Antibody (Pro1) is validated for use in Western Blotting and Immunocytochemistry and Dot Blot for the detection of dimethyl Histone H2B (Pro1).

Quality

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: 1 µg/mL of this antibody detected Histone H2B dimethylated at proline 1 in 10 µg of HeLa acid extract.

Target description

~17 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service