Z742667
2MAG steriMIXcontrol for use with steriMIXdrive
AC/DC input 100 V / 240 V AC, US 3-pin plug
Synonym(s):
control unit, power bank, power unit
All Photos(1)
About This Item
material
stainless steel
feature
20 W Stirring Power
packaging
pack of 1 ea
manufacturer/tradename
2mag 95200
parameter
0-40 °C temp. limit (at 80% humidity)
100-1200 rpm speed
W × D × H
200 mm × 175 mm × 48 mm
weight
1.8 kg
General description
Powerful control device for use with 2Mag steriMIXdrive stirring drives, with powerful 20 watt stirring power and mixWATCH stirring rod monitoring (without speed reduction). SoftStart function allows for reliable catching/centering and safe acceleration of the stirring rod, adjustable interval function for time-dependent restart, adjustable interval times for automatic restart of the stirrer after up to 240 seconds. Speed range from 100 to 1,200 rpm enables smooth stirring even at low speeds. Digital display for speed and interval setting, QuickSet for quickly setting the initial or maximum speed, QuickStop function to prevent overrunning and glass breakage, saving the last functional state when switching off.
Choose from one of the most recent versions:
Certificates of Analysis (COA)