Merck
GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX B-TA
GC Analysis of Alkylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ B-TA suitable for GC
材料
分析柱
产品编号
说明
价格
Astec ® CHIRALDEX B-TA 毛细管 GC 色谱柱
L × I.D. 30 m × 0.25 mm, df 0.12 μm
標準品
产品编号
说明
价格
2-氨基戊烷
97%
2-氨基庚烷
99%
(R)-(-)-2-氨基庚烷
99%
(S)-(+)-2-氨基庚烷
99%
CONDITIONS
column
Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 μm (73023AST)
gradient
80 °C, 2 °C/min to 110 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
0.6 μL, 80:1 split
sample
2.5 mg/mL each racemate in methanol (N-TFA derivatives)
说明
法律資訊
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC