返回
Nunc
®
EasyFill Cell Factory filter assembly cap
Products
制造商
包装大小
孔径(µm)
筛选产品
Showing 1-2 of 2
比较
货号
说明
价格
TMO140080
Nunc
®
EasyFill Cell Factory filter assembly cap,
high-density polyethylene, pore size 0.22 μm
展开
查看定价
TMO140067
Nunc
®
EasyFill Cell Factory filter assembly cap,
high-density polyethylene, pore size 1.0 μm