返回
Ace stopcock only, with inlet feed, for condenser "C" and "CR" assemblies
Products
制造商
筛选产品
Showing 1-2 of 2
比较
货号
说明
价格
Z682160
Ace stopcock only, with inlet feed, for condenser "C" and "CR" assemblies,
vented, fits Büchi
®
models R-200/R-205 and R-114/R-144
展开
查看定价
Z682179
Ace stopcock only, with inlet feed, for condenser "C" and "CR" assemblies,
hollow, fits Büchi
®
models R-200/R-205 and R-114/R-144