Merck
  • Acute, sub-acute and general pharmacological evaluation of 5-(3,4-methylenedioxyphenyl)-4-ethyl-2E,4E-pentadienoic acid piperidide (SK-20): a novel drug bioavailability enhancer.
Acute, sub-acute and general pharmacological evaluation of 5-(3,4-methylenedioxyphenyl)-4-ethyl-2E,4E-pentadienoic acid piperidide (SK-20): a novel drug bioavailability enhancer.
Environmental toxicology and pharmacology (2012-11-28)
Sheikh Rayees, Rohit Sharma, Gurdarshan Singh, Ishtiyaq Ahmad Najar, Amarinder Singh, Dastagir Basheer Ahamad, Subash Chander Sharma, Manoj Kumar Tikoo, Vijay Kumar Gupta, Payare Lal Sangwan, Surjeet Singh, Surinder Koul, Rakesh Kamal Johri
ABSTRACT
An efflux pump inhibitor, SK-20 (5-(3,4-methylenedioxyphenyle)-4 ethyl-2E,4E-pentadienoic acid piperidide), was assessed for its toxicity at three different pharmacological profiles: acute, sub-acute and general pharmacology with pharmacokinetics. In acute study, the SK-20 was found safe up to a dose of 2000 mg/kg (b.wt.); and at sub-acute, dosages of 50 and 100 mg/kg (b.wt.) were found to be safe. However, dosages of 200 mg or above per kg (b.wt.) showed some morphological alterations in cellular architecture of both liver and kidneys in both sexes, viz., mild vascular congestion along with sporadic hemorrhages and infiltration into renal and hepatic parenchyma by mononucleate cell. General pharmacological studies did not result into any alterations in analgesic, convulsions, rectal temperatures and in the rhythm or the rate of the intestinal motility or the secretion of the bile. While the respiratory and the cardiac rate remained normal, the only parameter to show was the blood pressure, which at all the doses tested, showed a tendency toward reduction. Characteristically, the SK-20 at all doses influenced pentobarbital-induced hypnosis positively and negatively to spontaneous motor activity in a dose dependent manner. Pharmacokinetics of SK-20 revealed it to have retention time at 10.2 min and half life 2.47 h.
Product Description
Supelco
Pentobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®