Skip to Content
HK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Yeast Extract
Synonym(s):
Select Yeast Extract, Yeast extract granulated, Yeast extract
CAS Number:
8013-01-2
EC Number:
232-387-9
Products
Product Category
extracts for media (7)
media supplements (1)
Brand
Millipore (7)
Sigma-Aldrich (4)
Biological Source
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Y1625
Yeast Extract,
for use in microbial growth medium
Expand
View Pricing
1.03753
Yeast Extract,
suitable for microbiology, medium granules
Expand
View Pricing
70161
Yeast Extract,
suitable for microbiology
Expand
View Pricing
92144
Yeast Extract,
suitable for microbiology
Expand
View Pricing
1.13885
Yeast Extract,
media component, suitable for microbiology
Expand
View Pricing
Y0875
Yeast Extract,
for use in microbial growth medium
Expand
View Pricing
09182
Yeast Extract,
for technical purposes (in fermentation)
Expand
View Pricing
Y1000
Yeast Extract,
powder, BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
Y0375
Yeast Extract,
powder
Expand
View Pricing
Y1626
Yeast Extract,
for use in microbial growth medium
Expand
View Pricing
89526
Yeast Extract,
micro-agglomerated
Expand
View Pricing