Skip to Content
Merck
All Photos(1)

Key Documents

90172

Supelco

1-Tridecanol

analytical standard

Synonym(s):

Tridecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12OH
CAS Number:
Molecular Weight:
200.36
Beilstein:
1739991
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

vapor density

6.9 (vs air)

Assay

≥96.5% (GC)

form

solid

shelf life

limited shelf life, expiry date on the label

color

, Colorless and White to Off White

bp

155-156 °C/15 mmHg (lit.)

mp

29-34 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCO

InChI

1S/C13H28O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14/h14H,2-13H2,1H3

InChI key

XFRVVPUIAFSTFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

250.0 °F - closed cup

Flash Point(C)

121.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service