Skip to Content
Merck
All Photos(6)

Key Documents

CWA-1114

Sigma-Aldrich

Anti-Human CD55 (143-30) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, (Sold Separately))

Synonym(s):

Complement decay-accelerating factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

143-30, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 41.4 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD55(1604)

General description

ColorWheelColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheelColorWheel® antibody with any ColorWheelColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheelColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheelColorWheel® technology requires both ColorWheelColorWheel® antibody and ColorWheelColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 143-30 is a mouse monoclonal antibody that detects human Complement decay-accelerating factor (CD55).

Immunogen

Phytohaemagglutinin (PHA) activated peripheral blood mononuclear cells.

Application

Quality Control Testing

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis:

Staining of one million HeLa cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1114, Anti-Human CD55 (143-30) ColorWheelColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheelColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-PE ColorWheelColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Tested Applications

Flow Cytometry Analysis: Staining of one million HeLa cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1114, Anti-Human CD55 (143-30) ColorWheelColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheelColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-APC ColorWheelColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Complement decay-accelerating factor (UniProt: P08174; also known as CD55) is encoded by the CD55 (also known as CR, DAF) gene (Gene ID: 1604) in human. CD55 is a phosphatidyl-inositol anchored glycoprotein that inhibits autologous complement cascade activation. It is expressed on the plasma membranes of all cell types that are in contact with plasma complement proteins. It is also found on the surfaces of epithelial cells lining extracellular compartments. Its variants are present in body fluids and in extracellular matrix. Seven different isoforms of CD55 have been described that are produced by alternative splicing. Isoforms 1 and 2 are membrane associated and isoforms 3 to 7 are secreted proteins. The major form of CD55 is a monomer; however, a minor non-disulfide-linked, covalent homodimeric form has also been reported. It is synthesized with a signal peptide (aa 1-34) and a propeptide (aa 354-381) that are subsequently cleaved off to generate the mature form (aa 35-353). It has four Sushi domains, and the first Sushi domain (SCR1) is not essential for its function, but SCR2 and 4 are required for the proper conformation for the active site on SCR3. CD55 recognizes C4b and C3b fragments that condense with cell-surface hydroxyl or amino groups when nascent C4b and C3b are locally generated during C4 and C3 activation. Its Interaction with cell-associated C4b and C3b polypeptides interferes with their ability to catalyze the conversion of C2 and factor B to enzymatically active C2a and Bb. This prevents the formation of C4b2a and C3bBb, the amplification convertases of the complement cascade. It also Inhibits complement activation by destabilizing and preventing the formation of C3 and C5 convertases, which prevents complement damage. Mutations in CD55 gene are known to cause complement hyperactivation and angiopathic thrombosis. Overexpression of CD55 has been reported in cases of HT29 colon carcinoma cells. (Ref.: Ozen, A., et al. (2017). N. Engl. J. Med. 377(1); 52-61; Ward, T., et al. (1994). EMBO J. 13(21); 5070-5074; Koretz, K., et al. (1992). Br. J. Cancer 66(5); 810-814).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service