Skip to Content
Merck
All Photos(1)

Key Documents

41768

Supelco

p-Toluic acid

analytical standard

Synonym(s):

4-Methylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein:
507600
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

274-275 °C (lit.)

mp

177-180 °C (lit.)

application(s)

pharmaceutical

format

neat

SMILES string

Cc1ccc(cc1)C(O)=O

InChI

1S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

LPNBBFKOUUSUDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

p-Toluic acid may be used as a reference standard for the determination of p-toluic acid in terephthalic acid by high performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

The purity is calculated based on the principle of mass balance and is mentioned on the CoA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

357.8 °F

Flash Point(C)

181 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 4-Carboxybenzaldehyde and p-Toluic Acid in Terephthalic Acid by High Performance Liquid Chromatography [J].
Delai P.
Petrochemical Technology, 7(1) (1995)
Mariana C F C B Damião et al.
Archiv der Pharmazie, 347(12), 885-895 (2014-10-07)
A novel class of benzo[d][1,3]dioxol-5-ylmethyl alkyl/aryl amide and ester analogues of capsaicin were designed, synthesized, and evaluated for their cytotoxic activity against human and murine cancer cell lines (B16F10, SK-MEL-28, NCI-H1299, NCI-H460, SK-BR-3, and MDA-MB-231) and human lung fibroblasts (MRC-5).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service