Skip to Content
Merck
All Photos(1)

Key Documents

AC113

Sigma-Aldrich

Anti-Neomycin Phosphotransferase II Antibody | AC113

from rabbit, purified by affinity chromatography

Synonym(s):

Aminoglycoside 3′-phosphotransferase, APH(3′)-II, APH(3′)II, Kanamycin kinase, type II, Neomycin-kanamycin phosphotransferase type II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Neomycin phosphotransferase II confers resistance to kanamycin and neomycin in bacteria and G418 (Geneticin®, G418 Sulfate) in mammalian cells. These antibiotics bind to ribosomal components and inhibit protein synthesis. NPT II inhibits these antibiotics through phosphorylation, which is thought to interfere with their active transport into the cell. Immunoblot analysis of transfected cell lysates with anti NPT II can be used to gauge relative transfection efficiencies in experiments where multiple transfections are performed in parallel with different plasmids.

Specificity

This antibody reacts to lysates from cells transfected or transformed with a plasmid containing a kanamycin/neomycin resistance gene, nptII (Aminoglycoside 3′-phosphotransferase).

Immunogen

Recombinant protein corresponding to full length neomycin phosphotransferase (NPT) II.

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Kinases & Phosphatases

Quality

Evaluated by Western Blot in untreated and neomycin treated HEK293 cells transfected with pUSEamp(-).

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Neomycin phosphotransferase II in 10 µg of neomycin treated HEK293 cells transfected with pUSEamp(-).

Target description

~29 kDa observed.

Physical form

Affinity purified
Hek 293 +/- transfection with pUSEamp(-)and neomycin treatment.
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and neomycin treated HEK293 cells transfected with pUSEamp(-).

Legal Information

Geneticin is a registered trademark of Gibco BRL Life Technologies, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saleh Bhar et al.
Human mutation, 41(11), 1918-1930 (2020-08-14)
Diamond-Blackfan anemia (DBA) is a ribosomopathy of variable expressivity and penetrance characterized by red cell aplasia, congenital anomalies, and predisposition to certain cancers, including early-onset colorectal cancer (CRC). DBA is primarily caused by a dominant mutation of a ribosomal protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service