technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
storage temp.
no temp limit
General description
for Prove 100, 300 and 600 Spectroquant®
This rugged case is ideal for transporting a Spectroquant® Prove 100, Prove 300, or Prove 600 (not included). It features a foam inlay that idealy protects the instrument. Therefore it can be used to transport the device from lab to lab or any other location if you for example conduct trainings and demonstrations for your colleagues.
This rugged case is ideal for transporting a Spectroquant® Prove 100, Prove 300, or Prove 600 (not included). It features a foam inlay that idealy protects the instrument. Therefore it can be used to transport the device from lab to lab or any other location if you for example conduct trainings and demonstrations for your colleagues.
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service