Merck
  • 5-Iodo-2-aminoindan (5-IAI): chemistry, pharmacology, and toxicology of a research chemical producing MDMA-like effects.
5-Iodo-2-aminoindan (5-IAI): chemistry, pharmacology, and toxicology of a research chemical producing MDMA-like effects.
Toxicology letters (2013-01-26)
M Coppola, R Mondola
ABSTRACT
In 2010, an internet snapshot of EMCDDA anticipated the presence of 5-iodo-2-aminoindan (5-IAI) within the recreational drug market. In 2011, this compound, a psychoactive derivative of 2-aminoindane, was identified in recreational products sold in the United Kingdom. 5-IAI is a rigid analogue of p-iodoamphetamine producing MDMA-like effects. The aim of this paper is to summarize the clinical, pharmacological, and toxicological information about this new potential drug of abuse.
Product Description
Supelco
(±)-MDA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®