Skip to Content
Merck
All Photos(1)

Key Documents

797-0020

GC Analysis of Alcohol Enantiomers on β-DEX 325

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: β-DEX 325, 30 m × 0.25 mm I.D., 0.25 μm (24308)
oven: 110 °C, 1-phenyl-2-propanol; 100 °C, isopinocampheol; 90 °C, 6-methyl-5-hepten-2-ol
inj. temp.: 220 °C
detector: FID, 300 °C
carrier gas: helium, 20 cm/sec @ analysis temperature
injection: 1 μL 100:1 split
sample: 1 mg/mL in methylene chloride

application(s)

chemicals and industrial polymers

Legal Information

DEX is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    6-Methyl-5-hepten-2-ol
    1-Phenyl-2-propanol, 98%

analytical column

Product No.
Description
Pricing

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service