22055-U
Swagelok® PTFE Ferrules
PTFE back ferrule, Swagelok for 1/4 in. tubing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
Recommended Products
material
PTFE back ferrule
packaging
pkg of 10 ea
I.D.
1/4 in.
fitting
Swagelok for 1/4 in. tubing
Looking for similar products? Visit Product Comparison Guide
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
Application
Swagelok® PTFE ferrules maybe used, for making all the connections between glass and PTFE present in the containment vessels, piping, and sampling equipment in a bench scale biofilter system, in an experimental study investigating the effects of periods of non-use on biofilter performance.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effect of periods of non-use on biofilter performance.
Jason MF and Loehr RC.
Journal of the Air & Waste Management Association (1995), 46 (6), 539-546 (1996)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service