Skip to Content
Merck
All Photos(4)

Key Documents

ZRB2816

Sigma-Aldrich

Anti-DKK1 Antibody, clone 1N16 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Dickkopf-1, Dkk-1, hDkk-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1N16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 28.67 kDa
observed mol wt ~30 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

porcine, horse, monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... DKK1(22943)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1N16 is a ZooMAb® recombinant rabbit monoclonal antibody that detects DKK1. It targets an epitope within 25 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal half of human DKK1.

Application

Quality Control Testing

Evaluated by Western Blotting in U-251 MG cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected DKK1 protein in U-251 MG cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound DKK1 peptide with at least one thousand-fold (1,000X) higher than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected DKK1 in human placenta tissue sections.

Immunocytochemistry Analysis : A 1:100 dilution from a representative lot detected DKK1 in PC-3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Dickkopf-related protein 1 (UniProt: O94907; also known as Dickkopf-1, Dkk-1, hDkk-1, SK) is encoded by the DKK1 (also known as UNQ492/PRO1008) gene (Gene ID: 22943) in human. Dickkopf-1 (DKK1) is a secreted protein that is a member of the Dickkopf family involved in modulating Wnt signaling pathways. DKK1 contains two cysteine-rich domains connected by a linker region, which are crucial for its function and interaction with other proteins, such as the Wnt co-receptors LRP5 and LRP6. DKK1 functions primarily as an antagonist of the Wnt signaling pathway, a key regulator of cell growth, differentiation, and embryonic development. By binding to LRP5/6, DKK1 prevents the formation of the Wnt-Frizzled-LRP complex, thereby inhibiting the canonical Wnt signaling pathway. This inhibition is critical for regulating processes such as bone formation and growth. DKK1 is expressed in a variety of tissues, with significant roles in both embryonic development and adult tissue homeostasis. Its expression is particularly noted in bone, where it is involved in the regulation of osteoblast and osteoclast activity, impacting bone density and turnover. Overexpression of DKK1 is associated with bone loss and osteoporosis, as it inhibits Wnt signaling which is crucial for bone formation. Conversely, reduced levels of DKK1 are seen in conditions characterized by excessive bone growth. DKK1 is also involved in cancer biology; it can either promote or inhibit tumor growth depending on the cancer type and the context of Wnt signaling activity. In some cancers, such as multiple myeloma, DKK1 expression is linked to poor prognosis due to its role in creating a bone-destructive environment around the tumor. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service