Key Documents
Z381462
Heating blocks for Techne® Dri-Block® heater
holds 12, 15 mm tubes
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
feature
Holds 12 x 15 mm tubes
manufacturer/tradename
Techne F3506
General description
Interchangeable blocks machined of aluminum for rapid heat transfer. Close tolerances assure efficient, rapid heating. Each block has a threaded thermometer hole for independent temperature monitoring, which also fits the block extraction tool.
Legal Information
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.