Skip to Content
Merck
All Photos(1)

Key Documents

T11100-HH

Sigma-Aldrich

DMT-dT Phosphoramidite

Synonym(s):

5′-O-(4,4′-dimethoxytrityl)-2′-deoxythymidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite], 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxythymidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-dT amidite, Thymidine, 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H49N4O8P
CAS Number:
Molecular Weight:
744.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

biological source

non-animal source

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder or granules

packaging

pkg of 1 × 1 kg T111012-01
pkg of 6 × 10 g (T111021-06)
pkg of 1 × 100 g (T111011-C02-01)
pkg of 6 × 20 g (T111020-06)
pkg of 1 × 500 g T111051-01

technique(s)

oligo synthesis: suitable

impurities

≤0.3 wt. % water content (Karl Fischer)

color

white to off-white

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxythymidine
base protecting group: none
5' protecting group: DMT
deprotection: fast/standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=C(C)C(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C40H49N4O8P/c1-27(2)44(28(3)4)53(50-23-11-22-41)52-35-24-37(43-25-29(5)38(45)42-39(43)46)51-36(35)26-49-40(30-12-9-8-10-13-30,31-14-18-33(47-6)19-15-31)32-16-20-34(48-7)21-17-32/h8-10,12-21,25,27-28,35-37H,11,23-24,26H2,1-7H3,(H,42,45,46)/t35-,36+,37+,53?/m0/s1

InChI key

UNOTXUFIWPRZJX-CEXSRUIHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service