Skip to Content
Merck
All Photos(1)

Key Documents

SML3742

GK13S

new

≥98% (HPLC)

Synonym(s):

(S)-1-Cyano-N-(1-(4-(pent-4-yn-1ylcarbamoyl)phenyl)-1H-imidazol-4-yl)pyrrolidine-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N6O2
Molecular Weight:
390.44
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Cell penetrant, potent and non-toxic activity-based probe for deubiquitinase UCHL1; chemogenomic probe suitable for the cellular study of UCHL1.


GK13S is a cell penetrant, potent and non-toxic activity-based probe for deubiquitinase UCHL1. GK13S, but not GK16S, led to the reduction of monoubiquitin levels in the human glioblastoma cell line U-87 MG. GK13S together with GK16S comprises a set of chemogenomic probes for UCHL1, which is suitable for investigation of cellular functions of UCHL. GK13S also binds to PARK7, C21orf33, ISOC1 andNIT2.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service