Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306277

Sigma-Aldrich

ANTI-OSTEOCALCIN(N-TERMINAL) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

BGLAP, Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, Osteocalcin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: 1:50-1:100
western blot: 1:1000
citations:
13

biological source

rabbit

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

10963 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BGLAP(632)

General description

Bone γ-carboxyglutamate protein (BGLAP) or osteocalcin (OCN) is a non-collagenous bone matrix protein expressed by osteoblasts. It possesses a Gla (γ-carboxyglutamate) domain. The gene encoding OCN is localized on human chromosome 1q22.

Application

ANTI-OSTEOCALCIN(N-TERMINAL) antibody produced in rabbit has been used in western blotting and immnuohistochemistry.

Biochem/physiol Actions

Bone γ-carboxyglutamate protein (BGLAP) regulates osteogenic differentiation of mesenchymal stromal cells. It has a role in bone turnover and mineralization. The protein also has a role in energy metabolism. BGLAP may act as a mediator between glucose metabolism and bone tissue.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen sulfide regulates bone remodeling and promotes orthodontic tooth movement
Haiya Pu, et al.
Molecular Medicine Reports (2017)
Evelin Carine Alves Silva et al.
Journal of endodontics, 46(10), 1470-1477 (2020-07-20)
Bio-C Sealer (BC; Angelus, Londrina, PR, Brazil) and Sealer Plus BC (SPBC; MK Life, Porto Alegre, Brazil) are new ready-to-use bioceramic endodontic sealers. The aim of this study was to evaluate the biocompatibility and bioactive potential of BC and SPBC
Osteocalcin Mediates Biomineralization during Osteogenic Maturation in Human Mesenchymal Stromal Cells
Yu-Tzu Tsao, et al.
International Journal of Molecular Sciences (2017)
Gregory R. Mundy and T. John Martin
Physiology and Pharmacology of Bone (2012)
Knockdown of Indian hedgehog protein induces an inhibition of cell growth and differentiation in osteoblast MC3T3-E1 cells
Ang Deng, et al.
Molecular Medicine Reports (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service