A7552
Liquid chromatography column fittings
Adapter, Luer Lock to 0.038 in. tubing (F)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
grade
for chromatography
technique(s)
LPLC: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Tubing adapter consists of a five foot length of PTFE tubing with integral CTFE Luer-Lock fitting.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service