60541
Potassium tetraborate tetrahydrate
purum p.a., ≥99.0% (T)
Synonym(s):
Potassium biborate
About This Item
Recommended Products
grade
purum p.a.
Assay
≥99.0% (T)
form
powder
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤5000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg
SMILES string
O.O.O.O.[K+].[K+].[O-]B([O-])OB1OB(OB=O)O1
InChI
1S/B4O7.2K.4H2O/c5-1-8-3-10-4(11-3)9-2(6)7;;;;;;/h;;;4*1H2/q-2;2*+1;;;;
InChI key
FZICZLCGJZRAHS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service