Skip to Content
Merck
All Photos(1)

Key Documents

69581

Supelco

Pyrrolidine

analytical standard

Synonym(s):

Tetrahydropyrrole, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
Beilstein:
102395
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.45 (vs air)

vapor pressure

128 mmHg ( 39 °C)
49 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

653 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.442-1.444
n20/D 1.443 (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C1CCNC1

InChI

1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2

InChI key

RWRDLPDLKQPQOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

V A Efimov et al.
Bioorganicheskaia khimiia, 36(6), 725-746 (2011-02-15)
In order to improve physicochemical and biological properties of natural oligonucleotides in particular increasing their affinity for nucleic acids, the selectivity of action and biological sustainability, several types of DNA mimics were designed. The survey collected data on the synthesis
Shao-Ju Chien et al.
BMC complementary and alternative medicine, 15, 61-61 (2015-04-19)
Homocysteine and pro-inflammatory mediators such as cyclooxygenase-2 (COX-2) have been linked to vascular dysfunction and risks of cardiovascular diseases. Fulvic acid (FA), a class of compounds of humic substances, possesses various pharmacological properties. However, the effect of FA on inflammatory
Rosa A Sierra et al.
Cancer immunology research, 2(8), 800-811 (2014-05-17)
An impaired antitumor immunity is found in patients with cancer and represents a major obstacle in the successful development of different forms of immunotherapy. Signaling through Notch receptors regulates the differentiation and function of many cell types, including immune cells.
Seok-Hwi Park et al.
Organic & biomolecular chemistry, 13(15), 4539-4550 (2015-03-18)
Concise and stereocontrolled syntheses of (+)-hyacinthacine A2 and sphingofungin B were achieved via a diastereomerically enriched oxazine intermediate. The key strategies include palladium(0)-catalyzed intramolecular oxazine formation and diastereoselective nucleophilic addition to an aldehyde. (+)-Hyacinthacine A2 was synthesized in 13 steps
Karen Menuz et al.
PLoS genetics, 10(11), e1004810-e1004810 (2014-11-21)
Many insect vectors of disease detect their hosts through olfactory cues, and thus it is of great interest to understand better how odors are encoded. However, little is known about the molecular underpinnings that support the unique function of coeloconic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service