Skip to Content
Merck
All Photos(1)

Key Documents

45442

Supelco

Diclofop-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14Cl2O4
CAS Number:
Molecular Weight:
341.19
Beilstein:
2224754
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

42-48 °C (if solid)

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(C)Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1

InChI

1S/C16H14Cl2O4/c1-10(16(19)20-2)21-12-4-6-13(7-5-12)22-15-8-3-11(17)9-14(15)18/h3-10H,1-2H3

InChI key

BACHBFVBHLGWSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diclofop-methyl is an early post-emergence phenoxypropanoic acid herbicide. Its mode of action involves inhibition of acetyl CoA carboxylase (ACCase) activity, which in turn decreases fatty acid synthesis in weeds and inhibits photosynthesis and the activity of meristem.

Application

Diclofop-methyl may be used as a reference standard for the determination of diclofop-methyl in soil samples by gas-liquid chromatography (GLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose L De Prado et al.
Journal of agricultural and food chemistry, 53(6), 2185-2191 (2005-03-17)
Three diclofop-methyl (DM) resistant biotypes of Lolium rigidum (R1, R2, and R3) were found in different winter wheat fields in Spain, continuously treated with DM, DM + chlortoluron, or DM + isoproturon. Herbicide rates that inhibited shoot growth by 50%
Vanesa Guillén-Casla et al.
Journal of agricultural and food chemistry, 56(7), 2303-2309 (2008-03-15)
Simple one- and two-dimensional high-performance liquid chromatography (HPLC) methods for the simultaneous enantiomeric determination of alkyloxyphenoxypropionic acid herbicides is presented. Compounds studied were ( R, S)-2-[4-(2,4-dichlorophenoxy)phenoxy]propionic acid (diclofop-acid) and ( R, S)-2-[4-(2,4-dichlorophenoxy)]methyl propionate (diclofop-methyl). Mobile phases necessary to separate their
Xiyun Cai et al.
Environmental toxicology and chemistry, 26(5), 970-975 (2007-05-25)
Information on the effects of pesticide degradation on aquatic organisms is needed to properly evaluate the ecotoxicity arising from the use of pesticides and for aquatic risk assessment. This work evaluated the toxicity of diclofop-methyl (DM) and its two major
Fatma Ünal et al.
Drug and chemical toxicology, 34(4), 390-395 (2011-07-01)
Diclofop-methyl (DM) is a chlorophenoxy derivative used in large quantities for the control of annual grasses in grain and vegetable crops. In this study, the genotoxic effects of DM were investigated by measuring chromosomal aberrations (CAs) in mouse bone-marrow cells
Qin Yu et al.
Planta, 230(4), 713-723 (2009-07-16)
This study investigates mechanisms of multiple resistance to glyphosate, acetyl-coenzyme A carboxylase (ACCase) and acetolactate synthase (ALS)-inhibiting herbicides in two Lolium rigidum populations from Australia. When treated with glyphosate, susceptible (S) plants accumulated 4- to 6-fold more shikimic acid than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service