Skip to Content
Merck
All Photos(1)

Key Documents

37068

Supelco

Chloride test sticks Quantofix®

500-3000 mg/L (chloride)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41113319
NACRES:
NA.21

measuring range

500-3000 mg/L (chloride)

application(s)

environmental
general analytical

storage temp.

2-8°C

General description

Contents:
100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (storage temperature 2–8 °C).

Gradation:
0 − 500 − 1000 − 1500 − 2000 − ≥3000 mg/L Cl-

Application

Quantofix® Chloride test strips may be used to remove zinc chloride from zinc sulphinate salts, during zinc bis(alkanesulphinate)s synthesis, which was intended to use as general reagent for formation of radical species.

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fionn O'Hara et al.
Nature protocols, 8(6), 1042-1047 (2013-05-04)
The present protocol details the synthesis of zinc bis(alkanesulfinate)s that can be used as general reagents for the formation of radical species. The zinc sulfinates described herein are generated from the corresponding sulfonyl chlorides by treatment with zinc dust. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service