TRNABAK-RO
Roche
tRNA
from bakers yeast
Synonym(s):
tRNA, transfer RNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
biological source
bakers yeast
form
lyophilized
packaging
pkg of 100 mg (10109495001)
pkg of 500 mg (10109509001)
manufacturer/tradename
Roche
storage temp.
−20°C
General description
Transfer RNAs (tRNA) are responsible for amino acid transport and the correct reading frame in protein biosynthesis. The knowledge of their tertiary structure and interactions with proteins and ribosomes is important for the understanding of mechanisms involved in protein biosynthesis.
Application
Transfer RNA (tRNA) has been used for the RNA pull down assay.
Quality
This product is tested for the absence of RNase according to the current Quality Control procedures.
1mg of dry substance corresponds to 16 A260 units.
1mg of dry substance corresponds to 16 A260 units.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jean-Baptiste Marq et al.
The Journal of biological chemistry, 286(8), 6108-6116 (2010-12-17)
Arenavirus RNA genomes are initiated by a "prime and realign" mechanism, such that the initiating GTP is found as a single unpaired (overhanging) nucleotide when the complementary genome ends anneal to form double-stranded (ds) RNA panhandle structures. dsRNAs modeled on
R Giegé et al.
Nucleic acids research, 26(22), 5017-5035 (1998-11-04)
Correct expression of the genetic code at translation is directly correlated with tRNA identity. This survey describes the molecular signals in tRNAs that trigger specific aminoacylations. For most tRNAs, determinants are located at the two distal extremities: the anticodon loop
Stéphanie Anchisi et al.
mBio, 6(2), e02349-e02349 (2015-03-05)
Many RNA viruses are detected by retinoic acid-inducible gene i (RIG-I), a cytoplasmic sensor that triggers an antiviral response upon binding non-self-RNA that contains a stretch of double-stranded RNA (dsRNA) bearing a base-paired 5' ppp nucleotide. To gain insight into
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service