XX1004707
1L Funnel, 47 mm, ground glass seal
Accessories for filter holders for sample preparation.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
diam.
47 mm
funnel capacity
1 L
application(s)
filtration
shipped in
ambient
General description
REMOVE FROM SCOPE
Application
Accessories for filter holders for sample preparation.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service