Skip to Content
Merck
All Photos(2)

Key Documents

MABN670

Sigma-Aldrich

Anti-LCN1 Antibody, clone 10B10

ascites fluid, clone 10B10, from mouse

Synonym(s):

TP, PMFA, Lipocalin-1, Tear lipocalin, Tlc, Tear prealbumin, Von Ebner gland protein, VEG protein, LCN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

10B10, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LCN1(3933)

General description

Lipocalin-1 or alternatively known as Tear lipocalin (Tlc), Tear prealbumin (TP) or Von Ebner gland protein (VEG) is encoded by the human gene named LCN1/VEGP and expressed in the salivary and lachrymal glands as well as prostate tissue, but recent research has also shown LCN1 to be produced by other glandular tissues such as the pituitary gland. Lipocalin-1 is part of a larger lipocalin protein family of extracellular proteins and shares the ability to bind hydrophobic ligands and lipids but LCN1 also has other abilities such as being an inhibitor of cysteine proteinases and thus plays a role in the control of inflammation. LCN1 also seems to be important in taste utilization and plays a role in the non-immunological defense against micro-organisms and viruses in the eye and mouth. EMD-Millipore’s Anti-LCN1 mouse monoclonal antibody has been tested in western blot against HEK293 transfected LCN1 lysates and in paraffin embedded immunohistochemistry on human prostate tissue sections and in fluorescent immunocytochemistry on transfected HEK293 cells in culture.

Immunogen

Purified recombinant fragment of human LCN1 expressed in E. Coli.

Application

Anti-LCN1 Antibody, clone 10B10 is a highly specific mouse monoclonal antibody, that targets Lipocalin & has been tested in western blotting, IHC & Immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected LCN1 in methanol-fixed HEK293 cells transfected with LCN1-hIgGFc.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected LCN1 in human prostate tissue.

Optimal working dilutions must be determined by end user.

Quality

Evaluated by Western Blotting in HEK293 and LCN1-hIgGFc transfected HEK293 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected LCN1-hIgGFc in LCN1-hIgGFc transfected HEK293 cell lysate.

Target description

~52 kDa observed. Uncharacterized bands may appear in some lysate(s).

Analysis Note

Control
HEK293 and LCN1-hIgGFc transfected HEK293 cell lysates

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service