Skip to Content
Merck
All Photos(1)

Key Documents

MABC1791

Sigma-Aldrich

Anti-hMENA11a Antibody, clone A1

Synonym(s):

ENAH, Protein enabled homolog, hMENA11a, Mammalian enabled variant 11a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A1, monoclonal

mol wt

calculated mol wt 66.5 kDa
observed mol wt ~90 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ENAH(55740)

Specificity

Clone A1 is a mouse monoclonal antibody that detects hMENA 11a. It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

A linear peptide corresponding to 18 amino acids from the C-terminal region of human MENA protein belonging to 11a exon.

Application

Quality Control Testing

Evaluated by Western Blotting in T47D cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected hMENA11a in T47D cell lysate.

Tested Applications

Immunocytochemistry Analysis: A representative lot detected MENA11a/ENAH in Western Blotting application (Di Modugno, F., et al. (2012). Proc Natl Acad Sci USA. 109(47):19280-5; Melchionna, R., et al. (2020). EMBO Rep. 21(11):e50078).

Western Blotting Analysis: A representative lot detected MENA11a/ENAH in Western Blotting applications (Di Modugno, F., et al. (2012). Proc Natl Acad Sci USA. 109(47):19280-5).

Immunohistochemistry Applications: A representative lot detected MENA11a/ENAH in Immunohistochemistry applications (Di Modugno, F., et al. (2012). Proc Natl Acad Sci USA. 109(47):19280-5).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein enabled homolog (UniProt: A0A097PIC4; also known as hMENA, Mammalian enabled variant 11a, ENAH) is encoded by the ENAH (also known as MENA) gene (Gene ID: 55740) in human. hMENA is a homotetrameric actin regulatory protein of the ENA/VASP family that plays a key role in cell processes that rely on actin cytoskeleton dynamics. Its expression is observed in a variety of tissues. It is overexpressed in a majority of breast cancer cell lines and primary breast tumor lesions. Its EVH2 domain mediates tetramerization, F-actin binding, and actin bundle formation. The EVH2 domain is comprised of three regions: Block A (aa 391-411) is a thymosin-like domain that is essential for G-actin binding; block B (aa 442-459) is required for F-actin binding and subcellular location, and block C (aa 554-588) is required for tetramerization. It also contains a KLKR motif (aa 400-403) within block A that is reported to be essential for G-actin binding and actin polymerization. hMENA is absent in normal breast tissue, but it is overexpressed in breast tumors, where its expression correlates with HER-2-positive, ER-negative, and high Ki 67 phenotype. Depletion of hMENA/ hMENA11 is reported to decrease HER3 phosphorylation, inhibits EGF- and NRG-mediated activation of EGFR and HER-2 and counteract growth factor-mediated cell proliferation. Silencing of hMENA/hMENA11a is reported to switch on molecules linked to apoptosis, including cleaved forms of poly (ADP-ribose) polymerase and caspase 9. A hMENA v6 variant of hMENA has also been described that increases cell invasion. In an isogenic model of human breast cancer progression, hMENA11a expression is observed in premalignant cells, whereas hMENA v6 expression is restricted to invasive cancer cells. (Ref.: Melchionna, R., et al. (2020). EMBO Rep. 21; e50078; Trono, P., et al. (2016). Mol. Cell. Oncol. 3(2); e1083648; Di Modugnoa, F., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(47); 19280-19285).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service