Skip to Content
Merck
All Photos(3)

Key Documents

ABN327

Sigma-Aldrich

Anti-SCRAPPER Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

F-box, Leucine rich repeat protein 20, FBXL20, FBl2, FBL20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FBXL20(84961)

General description

Members of the F-box protein family, such as SCRAPPER, are characterized by an approximately 40-amino acid F-box motif. SCRAPPER is selectively expressed in the brain, broadly expressed within the mouse CNS, and is abundant at the presynaptic membrane. It has orthologs in C. elegans, D. melanogaster, and mammals which suggests that it might function as an important membrane-localized E3 ligase in various species. SCRAPPER is a major presynaptic E3 ubiquitin ligase that acts through RIM1a via degradation and the ubiquitin-proteasome-system (UPS) pathway to critically regulate synaptic transmission. This identifies protein degradation as a mechanism for holding synaptic communication in check.

Immunogen

KLH-conjugated linear peptide corresponding to human SCRAPPER.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This SCRAPPER antibody is validated for use in WB & IHC for the detection of the SCRAPPER protein.
Western Blot Analysis: 1 µg/mL from a representative lot detected SCRAPPER in 10 µg/mL of RAW264.7 cell lysate.

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected SCRAPPER in human midbrain tissue.

Quality

Evaluated by Western Blot in human spleen tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected SCRAPPER in 10 µg/mL of human spleen tissue lysate.

Target description

~48 kDa observed. Uncharacterized bands may appear at ~52 kDa, ~58 kDa and ~70 kDa in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service