Skip to Content
Merck
All Photos(1)

Key Documents

65039-M

Sigma-Aldrich

Mollifex Tissue Softener

Softening agent for use with paraffin embedded tissues. Ideal for hard to section blocks and brittle tissues

Synonym(s):

Decalcifier

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116113

Quality Level

form

liquid

color

colorless

mp

-56.56 °C

transition temp

flash point 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

Application

Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Softening agent for use with paraffin embedded tissues. Ideal for hard to section blocks and brittle tissues.
Softening agent for use with paraffin embedded tissues. Ideal for hard to section blocks and brittle tissues

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service