Skip to Content
Merck
All Photos(1)

Key Documents

34875-M

Sigma-Aldrich

tert-Butyl methyl ether

suitable for HPLC, ≥99.8%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
55-56 °C (lit.)

grade

HPLC grade

vapor density

3.1 (vs air)

Assay

≥99.8%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.0005% peroxides (as H2O2)
≤0.002% free acid (as CH3COOH)
≤0.01% water (Karl Fischer)

color

APHA: ≤10

transmittance

220 nm, ≥40%
250 nm, ≥80%
260 nm, ≥90%
290 nm, ≥99%

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

UV absorption

λ: 210 nm Amax: ≤1.0
λ: 220 nm Amax: ≤0.40
λ: 250 nm Amax: ≤0.10
λ: 260 nm Amax: ≤0.05
λ: 300 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl ether (methyl tert-butyl ether, MTBE) is a gasoline additive. Its oxidative degradation by propane-oxidizing bacterial strains has been tested. Kinetic studies of its heat-assisted persulfate oxidation at various pH, temperature, oxidant concentration and ionic strength levels suggests that reaction exhibits a pseudo-first-order decay model.

Application

tert-Butyl methyl ether (methyl tert-butyl ether, MTBE) has been used in the isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC.

Packaging

Important notice
This product will be phased out towards early 2019. There is a replacement product available : 34875

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-27.4 °F - closed cup

Flash Point(C)

-33.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Chang Huang et al.
Chemosphere, 49(4), 413-420 (2002-10-09)
The kinetics of heat-assisted persulfate oxidation of methyl tert-butyl ether (MTBE) in aqueous solutions at various pH, temperature, oxidant concentration and ionic strength levels was studied. The MTBE degradation was found to follow a pseudo-first-order decay model. The pseudo-first-order rate
R J Steffan et al.
Applied and environmental microbiology, 63(11), 4216-4222 (1997-11-15)
Several propane-oxidizing bacteria were tested for their ability to degrade gasoline oxygenates, including methyl tert-butyl ether (MTBE), ethyl tert-butyl ether (ETBE), and tert-amyl methyl ether (TAME). Both a laboratory strain and natural isolates were able to degrade each compound after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service