Skip to Content
Merck
All Photos(1)

Key Documents

234

Sigma-Aldrich

Hematoxylin Powder (C.I. 75290)

HARLECO® Nuclear staining

Synonym(s):

Hematoxylin, H&E Stain, Gill

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
302.28
Colour Index Number:
75290
MDL number:
UNSPSC Code:
41116130
EC Index Number:
208-237-3

form

solid

mp

140 °C (Elimination of water of crystallisation)

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

room temp

SMILES string

O1C[C@@]2([C@H](c4c1c(c(cc4)O)O)c3c(cc(c(c3)O)O)C2)O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m1/s1

InChI key

WZUVPPKBWHMQCE-CJNGLKHVSA-N

General description

HARLECO® Hematoxylin
234-16 100g Glass Bottle

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used in various formulations of hematoxylin, such as Weigert, Harris, Gill, Mayer and Ehrlich.
HARLECO® Nuclear staining

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service