Z264776
Hydrion® Brilliant pH dip stiks
pH range 0.0-6.0
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41113034
NACRES:
NB.79
Recommended Products
feature
non-bleeding: no
refill: no
packaging
pkg of 100 ea
manufacturer/tradename
Hydrion
graduations and accuracy measuring range
accuracy: 0.5 pH unit
pH range
0.0-6.0
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
Direct reading pH stick with clear, bright color matches over the desired pH range.
Legal Information
Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service