Key Documents
WHA10370010
Whatman® glass microfiber filters with inorganic binder, Grade GF 6
diam. 185 mm
Synonym(s):
Z749893
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
feature
hydrophilic
packaging
pk of 100
manufacturer/tradename
Whatman 10370010
Whatman Article No. 28414750 (US reference)
diam.
185 mm
General description
Good retention for very fine particles. This filter is used in water pollution applications, for removing protein from difficult-to-filter beers, for determination of chlorophyll and phytoplankton residues, for the determination of filterable substances and the residue on ignition (dry weight), for the analysis of aggressive media (e.g., acidic gases), for scintillation measurements and for determination of the elemental iron content in the presence of iron oxides.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)