Key Documents
BR703720
BRAND® Transferpette® S pipette, adjustable volume, multi-channel
12-channel, volume 0.5-10 μL
Synonym(s):
Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Transferpette® S
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
12-channel
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
BRAND 703720
volume
0.5-10 μL
General description
The Transferpette S piston-operated multichannel pipettes are the perfect manual pipettes for the most demanding applications in the lab. They provide all of the features required by users working in the life sciences field. A particular advantage of the new multichannel pipette is the easy operation, e.g. when performing serial pipetting of immunological assays, while serial dilutions or when filling the 96-well plates for cell cultures.
The use of new innovative materials results in a pipette that is light, precise, rugged and reliable.
includes - Each Transferpette S -8/-12 pipette is DE-M marking and supplied with performance certificate, 2 x TipBox, filled with pipette tips from BRAND, 1 shelf/rack mount, 1 reagent reservoir, 1 mounting tool, silicone oil and 1 set of sealing rings made of FKM.
Features and Benefits
  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This video explains how to adjust the volume on the Transferpette before being ready to pipette.
This article and video demonstrate how to change the V-Rings and O-Rings on the tip cones of any Transferpette® Multichannel Pipette.
This article describes easy calibration techniques for mechanical and electronic liquid handling instruments from BRAND®.