Skip to Content
Merck
All Photos(1)

Key Documents

724696

Sigma-Aldrich

3-Aminobenzenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

141-143 °C

SMILES string

Nc1cccc(c1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-2-1-3-6(4-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

JPVKCHIPRSQDKL-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annabel Meyer et al.
PloS one, 8(9), e73536-e73536 (2013-09-17)
Understanding factors driving the ecology of N cycling microbial communities is of central importance for sustainable land use. In this study we report changes of abundance of denitrifiers, nitrifiers and nitrogen-fixing microorganisms (based on qPCR data for selected functional genes)
Tanzeela Abdul Fattah et al.
Bioorganic chemistry, 82, 123-128 (2018-10-13)
We describe the synthesis of a series of novel 1-aroyl/acyl-3-(3-aminosulfonylphenyl) thioureas (4a-k) acting as human carbonic anhydrase (hCA, EC 4.2.1.1) inhibitors. Reaction of alkyl/aryl isothiocyanates with 3-aminobenzenesulfonamide afforded a series of the title compounds incorporating a variety of short as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service