232645
2,6-Naphthalenedicarboxylic acid dipotassium salt
95%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
95%
form
solid
SMILES string
[K+].[K+].[O-]C(=O)c1ccc2cc(ccc2c1)C([O-])=O
InChI
1S/C12H8O4.2K/c13-11(14)9-3-1-7-5-10(12(15)16)4-2-8(7)6-9;;/h1-6H,(H,13,14)(H,15,16);;/q;2*+1/p-2
InChI key
QAYXDWGFSMUTBJ-UHFFFAOYSA-L
General description
Electrospray ionization mass spectrum of the potassium salt of 2,6-naphthalenedicarboxylic acid in a 3:1 water/methanol solution has been investigated.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Effects of organic acid pretreatment on the structure and pyrolysis reactivity of coals.
Fuel: The Science and Technology of Fuel and Energy, 80(2), 273-281 (2001)
Formation of gas-phase dianions and distonic ions as a general method for the synthesis of protected reactive intermediates. Energetics of 2, 3-and 2, 6-dehydronaphthalene.
Journal of the American Chemical Society, 122(43), 10689-10696 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service