Principaux documents
TEM-LC325AU
Sigma-Aldrich
Lacey Carbon Supported Gold Grids
size 300 mesh, size 50 nm , hole size, box of 25 ×
Synonyme(s) :
Lacey Carbon Support Film, 300 mesh Au Grids, 25pk
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25 ×
Taille
300 mesh
50 nm , hole size
Description générale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.