Principaux documents
SLW1517/01
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 25 mL
Synonyme(s) :
Pyrex, SciLabware 1517/01
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Capacité:
25 mL
Diam. × H:
36 mm × 70 mm
Matériaux:
borosilicate glass bottle
round bottle
white graduation
Matériaux
borosilicate glass bottle
round bottle
white graduation
Joint
Joints threaded neck (GL25)
Caractéristiques
graduations
without cap
with printed trace code
without pouring ring
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 1517/01
Capacité
25 mL
Diam. × H
36 mm × 70 mm
Description générale
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Informations légales
PYREX is a registered trademark of Corning, Inc.