Key Documents
CAD4173
Stainless steel 304 syringe needle
gauge 22, L 1 1/2 in.
About This Item
Code UNSPSC :
42142600
Nomenclature NACRES :
NB.03
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Cadence Science 4173
Longueur
1 1/2 in.
Calibre
22
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing