Skip to Content
Merck
All Photos(2)

Key Documents

V800443

Sigma-Aldrich

Water

suitable for HPLC, Vetec

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
technique(s):
HPLC: suitable
bp:
100 °C (lit.)
vapor pressure:
3 mmHg

Product Name

Water, suitable for HPLC

vapor density

<1 (vs air)

Quality Level

vapor pressure

3 mmHg

product line

Vetec

form

liquid

technique(s)

HPLC: suitable

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philippe Robert et al.
Investigative radiology, 50(8), 473-480 (2015-06-25)
To prospectively compare in healthy rats the effect of multiple injections of macrocyclic (gadoterate meglumine) and linear (gadodiamide) gadolinium-based contrast agents (GBCAs) on T1-weighted signal intensity in the deep cerebellar nuclei (DCN), including the dentate nucleus. Healthy rats (n =
Katrin Linke et al.
Biochimica et biophysica acta, 1837(1), 14-32 (2013-08-28)
Water is clearly important for the functioning of Photosystem II (PSII). Apart from being the very substrate that needs to be transported in this water oxidation enzyme, water is also vital for the transport of protons to and from the
Liming Zhou et al.
Nature, 509(7498), 86-90 (2014-04-25)
Tropical forests are global epicentres of biodiversity and important modulators of climate change, and are mainly constrained by rainfall patterns. The severe short-term droughts that occurred recently in Amazonia have drawn attention to the vulnerability of tropical forests to climatic
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service