Skip to Content
Merck
All Photos(1)

Key Documents

SRP8056

Sigma-Aldrich

Tim-3 (mouse): FC (human)

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE)

Synonym(s):

HAVcr-2, T-cell immunoglobulin and mucin domain-containing protein 3, TIM3, TIMD3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in CHO cells

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

monomer 45 kDa by calculation

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.06 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Havcr2(171285)

General description

Tim-3 is a type I transmembrane protein. It contains an immunoglobulin and a mucin-like domain in its extracellular portion and a tyrosine phosphorylation motif in its cytoplasmic portion. TIM-3 is preferentially expressed on Type 1 T-helper (Th1) and Type 1 CD8+ T (Tc1) cells. TIM-3 is also expressed on some dendritic cells. The gene encoding this protein is localized on mouse chromosome 11B1.1.

Biochem/physiol Actions

The T cell/transmembrane, immunoglobulin and mucin (TIM) family plays a crucial role in regulating immune responses, such as allergy, asthma, transplant tolerance, autoimmunity and also has a control over the response to viral infections. The unique structure of TIM immunoglobulin variable region domains allows highly specific recognition of phosphatidylserine (PtdSer), exposed on the surface of apoptotic cells. Tim-3 generates an inhibitory signal resulting in apoptosis of Type 1 T-helper (Th1) and Type 1 CD8+ T (Tc1) cells. It can also mediate phagocytosis of apoptotic cells and cross-presentation of antigen. It functions to inhibit aggressive Th1-mediated auto- and allo-immune responses. Tim-3 pathway blockade by administration of Tim-3: Fc fusion protein accelerates diabetes in non-obese diabetic mice, causes hyperproliferation of Th1 cells and Th1 cytokine release in an experimental autoimmune encephalomyelitis (EAE) model and prevents acquisition of transplantation tolerance induced by costimulation blockade.

Physical form

Lyophilized from 0.2 μm-filtered solution in PBS.

Reconstitution

Reconstitute at 100 μg/mL in sterile PBS.

Other Notes

The extracellular domain of mouse Tim-3 (aa 21-189) is fused to the N-terminus of the Fc region of human IgG1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TIM-3 is Expressed on Activated Human CD4+ T Cells and Regulates Th1 and Th17 Cytokines
William D. Hastings
European Journal of Immunology, 39(9), 2492-2501 (2009)
TIM Proteins and Immunity
Lawrence P. Kane
Journal of Immunology, 184(6), 2743-2749 (2010)
T and B cell hyperactivity and autoimmunity associated with niche-specific defects in apoptotic body clearance in TIM-4-deficient mice
Roselyn R
Proceedings of the National Academy of Sciences of the USA, 107.19, 8706-8711 (2010)
TIM-1 and TIM-3 Proteins in Immune Regulation
Ee Wern Su
Cytokine, 44(1), 9-13 (2008)
TIM genes: a family of cell surface phosphatidylserine receptors that regulate innate and adaptive immunity
Gordon J. Freeman
Immunological Reviews, 235(1), 172-189 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service