Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702774

Sigma-Aldrich

Anti-GSTK1 Antibody, clone 7J5B3, Rabbit Monoclonal

Synonym(s):

GST, GST 13-13, GST13, GST13-13, GSTK1-1, hGSTK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
7J5B3, monoclonal
application:
IF
WB
species reactivity:
mouse, mouse
technique(s):
immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000
citations:

biological source

rabbit

Quality Level

material

colorless

clone

7J5B3, monoclonal

form

liquid

mol wt

25 kDa

species reactivity

mouse, mouse

concentration

1.68 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MGPLPRTVELFYDVLSPYSWLGFEILCRYQNIWNINLQLRPSLITGIMKDSGNKPPGLLPRKGLYMANDLKLLRHHLQIPIHFPKDFLSVMLEKGSLSAM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTK1(373156)

General description

This gene encodes a member of the kappa class of the glutathione transferase superfamily of enzymes that function in cellular detoxification. The encoded protein is localized to the peroxisome and catalyzes the conjugation of glutathione to a wide range of hydrophobic substates facilitating the removal of these compounds from cells. Alternative splicing results in multiple transcript variants.[provided by RefSeq, Jan 2009]

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human GSTK1 (Q9Y2Q3).

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service